Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Theaflavine-3,3'-digallate 98%

Supplier: AMBEED, INC
Warning

A542803-1MG A542803-5MG
76649-686EA 23 USD InStock
76649-686 76649-688
Theaflavine-3,3'-digallate 98%
Theaflavine-3,3'-digallate
Precaución: for Research Use Only


Formula: C₄₃H₃₂O₂₀
MDL Number: MFCD06656329
CAS Number: 33377-72-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty