Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Isopropylamino)-3-nitrobenzonitrile 98%

Supplier: AMBEED, INC
Warning

A549645-1G A549645-250MG A549645-5G
76649-714EA 36 USD InStock
76649-714 76649-716 76649-718
4-(Isopropylamino)-3-nitrobenzonitrile 98%
4-(Isopropylamino)-3-nitrobenzonitrile
Precaución: for Research Use Only


Formula: C₁₀H₁₁N₃O₂
MDL Number: MFCD01570606
CAS Number: 355022-17-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty