Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrido[3,4-b]pyrazine-2,3(1H,4H)-dione 95%

Supplier: AMBEED, INC
Warning

A560945-100MG A560945-1G A560945-250MG
76649-794EA 5 USD InStock
76649-794 76649-796 76649-798
Pyrido[3,4-b]pyrazine-2,3(1H,4H)-dione 95%
Pyrido[3,4-b]pyrazine-2,3(1H,4H)-dione
Precaución: for Research Use Only


Formula: C₇H₅N₃O₂
MDL Number: MFCD15146306
CAS Number: 35251-84-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty