Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

WM-1119 99+%

Supplier: AMBEED, INC
Warning

A573581-100MG A573581-10MG A573581-1MG A573581-25MG A573581-50MG A573581-5MG
76649-894EA 154 USD InStock
76649-894 76649-896 76649-898 76649-900 76649-902 76649-904
WM-1119 99+%
WM-1119
Precaución: for Research Use Only


Formula: C₁₈H₁₃F₂N₃O₃S
MDL Number: MFCD31690337
CAS Number: 2055397-28-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty