Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-(Trifluoromethyl)oxazol-2-yl)-9H-xanthene-9-carboxamide 95%

Supplier: AMBEED, INC
Warning

A594240-100MG A594240-250MG A594240-50MG
76650-016EA 145 USD InStock
76650-016 77749-225 77749-226
N-(4-(Trifluoromethyl)oxazol-2-yl)-9H-xanthene-9-carboxamide 95%
N-(4-(Trifluoromethyl)oxazol-2-yl)-9H-xanthene-9-carboxamide
Precaución: for Research Use Only


Formula: C₁₈H₁₁F₃N₂O₃
MDL Number: MFCD31692711
CAS Number: 714971-87-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty