Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(2,5-dioxopyrrolidin-1-yl) heptanedioate 95%

Supplier: AMBEED, INC
Warning

A597741-1G A597741-250MG A597741-5G
76650-038EA 95 USD InStock
76650-038 76650-040 77622-384
Bis(2,5-dioxopyrrolidin-1-yl) heptanedioate 95%
Bis(2,5-dioxopyrrolidin-1-yl) heptanedioate
Precaución: for Research Use Only


Formula: C₁₅H₁₈N₂O₈
MDL Number: MFCD01318519
CAS Number: 74648-14-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty