Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Diamino-6-hydroxypyrimidine 97%

Supplier: AMBEED, INC
Warning

A608015-1000G A608015-100G A608015-500G A608015-5G
76650-138EA 98 USD InStock
76650-138 76650-140 76650-142 76650-144
2,4-Diamino-6-hydroxypyrimidine 97%
2,4-Diamino-6-hydroxypyrimidine
Precaución: for Research Use Only


Formula: C₄H₆N₄O
MDL Number: MFCD00006098
CAS Number: 56-06-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty