Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

MC-Val-Cit-PAB-OH 99%

Supplier: AMBEED, INC
Warning

A608926-100MG A608926-10MG A608926-1MG A608926-250MG A608926-50MG A608926-5MG A608926-5G
76650-164EA 88 USD InStock
76650-164 76650-166 76650-168 76650-170 76650-172 76650-174 77622-388
MC-Val-Cit-PAB-OH 99%
MC-Val-Cit-PAB-OH
Precaución: for Research Use Only


Formula: C₂₈H₄₀N₆O₇
MDL Number: MFCD26142966
CAS Number: 159857-80-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty