Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 6-hydroxynaphthalene-2-sulfonate xhydrate 97%

Supplier: AMBEED, INC
Warning

A613791-1000G A613791-500G
76650-224EA 39 USD InStock
76650-224 76650-226
Sodium 6-hydroxynaphthalene-2-sulfonate xhydrate 97%
Sodium 6-hydroxynaphthalene-2-sulfonate xhydrate
Precaución: for Research Use Only


Formula: C₁₀H₉NaO₅S
MDL Number: MFCD10565653
CAS Number: 825652-02-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty