Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nidufexor 95%

Supplier: AMBEED, INC
Warning

A615253-10MG A615253-1MG A615253-5MG
76650-228EA 42 USD InStock
76650-228 76650-230 76650-232
Nidufexor 95%
Nidufexor
Precaución: for Research Use Only


Formula: C₂₇H₂₂ClN₃O₄
MDL Number: MFCD31657267
CAS Number: 1773489-72-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty