Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 6-chloropyridine-2-sulfinate 95%

Supplier: AMBEED, INC
Warning

A618451-100MG A618451-1G A618451-250MG
76650-252EA 91 USD InStock
76650-252 76650-254 76650-256
Sodium 6-chloropyridine-2-sulfinate 95%
Sodium 6-chloropyridine-2-sulfinate
Precaución: for Research Use Only


Formula: C₅H₃ClNNaO₂S
MDL Number: MFCD30536977
CAS Number: 2682112-92-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty