Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-(tert-butyl)benzo[b]thiophene 97%

Supplier: AMBEED, INC
Warning

A1349684-1G A1349684-250MG A1349684-5G A1349684-25G
76658-116EA 10 USD InStock
76658-116 76658-118 76658-120 77621-626
3-Bromo-5-(tert-butyl)benzo[b]thiophene 97%
3-Bromo-5-(tert-butyl)benzo[b]thiophene
Precaución: for Research Use Only


Formula: C₁₂H₁₃BrS
MDL Number: MFCD28593666
CAS Number: 1780644-81-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty