Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (4-(oxazol-5-yl)benzyl)carbamate 97%

Supplier: AMBEED, INC
Danger

A629590-1G A629590-250MG
76650-352EA 51 USD InStock
76650-352 76650-354
tert-Butyl (4-(oxazol-5-yl)benzyl)carbamate 97%
tert-Butyl (4-(oxazol-5-yl)benzyl)carbamate
Precaución: for Research Use Only


Formula: C₁₅H₁₈N₂O₃
MDL Number: MFCD28965257
CAS Number: 1360616-36-9
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty