Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Fmoc-N'-methyl-L-asparagine 97%

Supplier: AMBEED, INC
Warning

A634766-100MG A634766-50MG A634766-5G
76650-440EA 41 USD InStock
76650-440 76650-442 76650-444
N-Fmoc-N'-methyl-L-asparagine 97%
N-Fmoc-N'-methyl-L-asparagine
Precaución: For research use only.


Formula: C₂₀H₂₀N₂O₅
MW: 368.3832 g/mol
MDL Number: MFCD30533782
CAS Number: 149204-93-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty