Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

t-Boc-N-Amido-PEG9-Amine 98%

Supplier: AMBEED, INC
Warning

A642480-100MG A642480-1G A642480-250MG
76650-534EA 34 USD InStock
76650-534 76650-536 76650-538
t-Boc-N-Amido-PEG9-Amine 98%
t-Boc-N-Amido-PEG9-Amine
Precaución: for Research Use Only


Formula: C₂₅H₅₂N₂O₁₁
MDL Number: MFCD03453249
CAS Number: 890091-43-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty