Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N'-1,2-Ethanediyl bis-hexadecanamide 95%

Supplier: AMBEED, INC
Warning

A642822-100MG A642822-250MG
76650-548EA 114 USD InStock
76650-548 76650-550
N,N'-1,2-Ethanediyl bis-hexadecanamide 95%
N,N'-1,2-Ethanediyl bis-hexadecanamide
Precaución: for Research Use Only


Formula: C₃₄H₆₈N₂O₂
MW: 536.9159 g/mol
MDL Number: MFCD19705254
CAS Number: 5518-18-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty