Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dodecaethylene glycol monomethyl ether 95%

Supplier: AMBEED, INC
Warning

A657004-100MG A657004-1G A657004-250MG A657004-5G
76650-676EA 11 USD InStock
76650-676 76650-678 76650-680 77573-116
Dodecaethylene glycol monomethyl ether 95%
Dodecaethylene glycol monomethyl ether
Precaución: for Research Use Only


Formula: C₂₅H₅₂O₁₃
MDL Number: MFCD06797088
CAS Number: 5702-16-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty