Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,3,6-Trichlorophenyl)methanamine 95%

Supplier: AMBEED, INC
Warning

A664373-1G A664373-250MG
76650-734EA 354 USD InStock
76650-734 77749-368
(2,3,6-Trichlorophenyl)methanamine 95%
(2,3,6-Trichlorophenyl)methanamine
Precaución: for Research Use Only


Formula: C₇H₆Cl₃N
MDL Number: MFCD07786738
CAS Number: 4960-49-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty