Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Benzyl-(9Z,12Z)-octadecadienamide 98%

Supplier: AMBEED, INC
Warning

A667149-10MG A667149-1MG A667149-25MG A667149-5MG
76650-754EA 29 USD InStock
76650-754 76650-756 76650-758 76650-760
N-Benzyl-(9Z,12Z)-octadecadienamide 98%
N-Benzyl-(9Z,12Z)-octadecadienamide
Precaución: for Research Use Only


Formula: C₂₅H₃₉NO
MDL Number: MFCD30723181
CAS Number: 18286-71-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty