Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Isopropylphenyl isocyanate 97%

Supplier: AMBEED, INC
Danger

A667634-10G A667634-1G A667634-5G
76650-774EA 136 USD InStock
76650-774 76650-776 76650-778
2-Isopropylphenyl isocyanate 97%
2-Isopropylphenyl isocyanate
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO
MDL Number: MFCD00013850
CAS Number: 56309-56-9
UN: 2206
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty