Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Chlorophenyl)prop-2-yn-1-one 97%

Supplier: AMBEED, INC
Warning

A673088-100MG A673088-1G A673088-250MG
76650-838EA 64 USD InStock
76650-838 76650-840 76650-842
1-(2-Chlorophenyl)prop-2-yn-1-one 97%
1-(2-Chlorophenyl)prop-2-yn-1-one
Precaución: for Research Use Only


Formula: C₉H₅ClO
MDL Number: MFCD21263012
CAS Number: 3463-33-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty