Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Boc-1-piperazinium-1-ylmethyl)trifluoroborate internal salt 98%

Supplier: AMBEED, INC
Warning

A674923-1G A674923-5G
76650-896EA 38 USD InStock
76650-896 76650-898
(4-Boc-1-piperazinium-1-ylmethyl)trifluoroborate internal salt 98%
(4-Boc-1-piperazinium-1-ylmethyl)trifluoroborate internal salt
Precaución: for Research Use Only


Formula: C₁₀H₂₀BF₃N₂O₂
MW: 268.0842 g/mol
MDL Number: MFCD20134170
CAS Number: 1268340-97-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty