Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3,5-Dichloropyrazin-2-yl)methanamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A681539-100MG A681539-250MG A681539-50MG A681539-1G
76650-982EA 46 USD InStock
76650-982 76650-984 76650-986 77749-404
(3,5-Dichloropyrazin-2-yl)methanamine hydrochloride 95%
(3,5-Dichloropyrazin-2-yl)methanamine hydrochloride
Precaución: for Research Use Only


Formula: C₅H₆Cl₃N₃
MDL Number: MFCD29761013
CAS Number: 2089319-25-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty