Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-(cyclohexyloxy)benzene 97%

Supplier: AMBEED, INC
Warning

A688231-1G A688231-250MG A688231-25G A688231-5G
76651-068EA 62 USD InStock
76651-068 76651-070 76651-072 76651-074
1-Bromo-4-(cyclohexyloxy)benzene 97%
1-Bromo-4-(cyclohexyloxy)benzene
Precaución: for Research Use Only


Formula: C₁₂H₁₅BrO
MDL Number: MFCD11114686
CAS Number: 30752-31-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty