Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,5,6-Tetra(9H-carbazol-9-yl)-1,3-Dicyanobenzene 97%

Supplier: AMBEED, INC
Warning

A689725-100MG A689725-250MG A689725-50MG A689725-1G
76651-088EA 10 USD InStock
76651-088 76651-090 76651-092 77552-096
2,4,5,6-Tetra(9H-carbazol-9-yl)-1,3-Dicyanobenzene 97%
2,4,5,6-Tetra(9H-carbazol-9-yl)-1,3-Dicyanobenzene
Precaución: for Research Use Only


Formula: C₅₆H₃₂N₆
MDL Number: MFCD27939633
CAS Number: 1416881-52-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty