Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Iodo-1H-pyrazole 95%

Supplier: AMBEED, INC
Warning

A696189-10G A696189-1G A696189-250MG A696189-25G A696189-5G
76651-144EA 138 USD InStock
76651-144 76651-146 76651-148 76651-150 76651-152
5-Iodo-1H-pyrazole 95%
5-Iodo-1H-pyrazole
Precaución: for Research Use Only


Formula: C₃H₃IN₂
MDL Number: MFCD01463487
CAS Number: 1007351-17-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty