Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

22-Bromo-2,5,8,11,14,17,20-heptaoxadocosane 95%

Supplier: AMBEED, INC
Warning

A698958-100MG A698958-250MG
76651-184EA 42 USD InStock
76651-184 76651-186
22-Bromo-2,5,8,11,14,17,20-heptaoxadocosane 95%
22-Bromo-2,5,8,11,14,17,20-heptaoxadocosane
Precaución: for Research Use Only


Formula: C₁₅H₃₁BrO₇
MDL Number: MFCD29060064
CAS Number: 104518-25-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty