Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

20-Bromo-3,6,9,12,15,18-hexaoxaicosan-1-ol 95%

Supplier: AMBEED, INC
Warning

A700471-1G A700471-100MG
76651-204EA 200 USD InStock
76651-204 77622-498
20-Bromo-3,6,9,12,15,18-hexaoxaicosan-1-ol 95%
20-Bromo-3,6,9,12,15,18-hexaoxaicosan-1-ol
Precaución: for Research Use Only


Formula: C₁₄H₂₉BrO₇
MDL Number: MFCD31656971
CAS Number: 86220-35-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty