Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N'-(6,7-Dihydroquinolin-8(5H)-ylidene)-4-(pyridin-2-yl)piperazine-1-carbothiohydrazide 98%

Supplier: AMBEED, INC
Warning

A703883-100MG A703883-10MG A703883-1MG A703883-25MG A703883-50MG A703883-5MG
76651-232EA 347 USD InStock
76651-232 76651-234 76651-236 76651-238 76651-240 76651-242
N'-(6,7-Dihydroquinolin-8(5H)-ylidene)-4-(pyridin-2-yl)piperazine-1-carbothiohydrazide 98%
N'-(6,7-Dihydroquinolin-8(5H)-ylidene)-4-(pyridin-2-yl)piperazine-1-carbothiohydrazide
Precaución: for Research Use Only


Formula: C₁₉H₂₂N₆S
MDL Number: MFCD28502211
CAS Number: 1039455-84-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty