Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Bis(4-bromophenyl)-2,4,6-trimethylaniline 98% (by GC)

Supplier: AMBEED, INC
Warning

A708868-100MG A708868-1G A708868-250MG
76651-308EA 26 USD InStock
76651-308 76651-310 76651-312
N,N-Bis(4-bromophenyl)-2,4,6-trimethylaniline 98% (by GC)
N,N-Bis(4-bromophenyl)-2,4,6-trimethylaniline
Precaución: for Research Use Only


Formula: C₂₁H₁₉Br₂N
MW: 445.20 g/mol
MDL Number: MFCD30534245
CAS Number: 663943-27-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty