Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(2-Methylbenzo[d]thiazol-6-yl)-2,3-diphenyl-5-(pyridin-2-ylamino)pyrazolo[1,5-a]pyrimidin-7(4H)-one ≥98%

Supplier: AMBEED, INC
Warning

A711197-10MG A711197-1MG A711197-25MG A711197-5MG
76651-354EA 407 USD InStock
76651-354 76651-356 76651-358 76651-360
6-(2-Methylbenzo[d]thiazol-6-yl)-2,3-diphenyl-5-(pyridin-2-ylamino)pyrazolo[1,5-a]pyrimidin-7(4H)-one ≥98%
6-(2-Methylbenzo[d]thiazol-6-yl)-2,3-diphenyl-5-(pyridin-2-ylamino)pyrazolo[1,5-a]pyrimidin-7(4H)-one
Precaución: for Research Use Only


Formula: C₃₁H₂₂N₆OS
MDL Number: MFCD31657415
CAS Number: 2201057-10-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty