Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diisopropyl sulfate 97%

Supplier: AMBEED, INC
Danger

A712832-250MG A712832-5G
76651-382EA 13 USD InStock
76651-382 76651-384
Diisopropyl sulfate 97%
Diisopropyl sulfate
Precaución: for Research Use Only


Formula: C₆H₁₄O₄S
MW: 182.23 g/mol
MDL Number: MFCD01682898
CAS Number: 2973-10-6
UN: 1760
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty