Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-(4-Amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)phenyl)-3-(2-fluoro-5-(trifluoromethyl)phenyl)urea 99+%

Supplier: AMBEED, INC
Warning

A712840-100MG A712840-10MG A712840-1MG A712840-25MG A712840-5MG A712840-50MG
76651-386EA 95 USD InStock
76651-386 76651-388 76651-390 76651-392 76651-394 77622-534
1-(4-(4-Amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)phenyl)-3-(2-fluoro-5-(trifluoromethyl)phenyl)urea 99+%
1-(4-(4-Amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)phenyl)-3-(2-fluoro-5-(trifluoromethyl)phenyl)urea
Precaución: for Research Use Only


Formula: C₂₂H₁₉F₄N₇O
MDL Number: MFCD31382193
CAS Number: 1384071-99-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty