Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dibromothioanisole 97%

Supplier: AMBEED, INC
Warning

A714490-1G A714490-250MG A714490-5G
76651-430EA 73 USD InStock
76651-430 76651-432 76651-434
2,5-Dibromothioanisole 97%
2,5-Dibromothioanisole
Precaución: for Research Use Only


Formula: C₇H₆Br₂S
MDL Number: MFCD13181616
CAS Number: 134646-03-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty