Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-Alanine methylamide hydrochloride 95%

Supplier: AMBEED, INC
Warning

A716215-10G A716215-1G A716215-5G
76651-456EA 452 USD InStock
76651-456 76651-458 76651-460
D-Alanine methylamide hydrochloride 95%
D-Alanine methylamide hydrochloride
Precaución: for Research Use Only


Formula: C₄H₁₁ClN₂O
MDL Number: MFCD22056408
CAS Number: 61302-99-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty