Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-7-fluoro-2-(trifluoromethyl)-1H-benzo[d]imidazole 95%

Supplier: AMBEED, INC
Warning

A719038-100MG A719038-1G A719038-250MG
76651-518EA 32 USD InStock
76651-518 76651-520 76651-522
5-Bromo-7-fluoro-2-(trifluoromethyl)-1H-benzo[d]imidazole 95%
5-Bromo-7-fluoro-2-(trifluoromethyl)-1H-benzo[d]imidazole
Precaución: for Research Use Only


Formula: C₈H₃BrF₄N₂
MW: 283.0204 g/mol
MDL Number: MFCD31698991
CAS Number: 2089920-05-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty