Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-4,4-Difluoropyrrolidine-2-carbonitrile hydrochloride 95%

Supplier: AMBEED, INC
Warning

A720519-100MG A720519-1G A720519-250MG A720519-5G
76651-544EA 34 USD InStock
76651-544 76651-546 76651-548 77622-558
(S)-4,4-Difluoropyrrolidine-2-carbonitrile hydrochloride 95%
(S)-4,4-Difluoropyrrolidine-2-carbonitrile hydrochloride
Precaución: for Research Use Only


Formula: C₅H₇ClF₂N₂
MW: 168.5723 g/mol
MDL Number: MFCD28403267
CAS Number: 869489-04-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty