Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ML-385 97%

Supplier: AMBEED, INC
Warning

A722849-10MG A722849-1MG A722849-25MG A722849-5MG
76651-594EA 58 USD InStock
76651-594 76651-596 76651-598 76651-600
ML-385 97%
ML-385
Precaución: for Research Use Only


Formula: C₂₉H₂₅N₃O₄S
MDL Number: MFCD06616124
CAS Number: 846557-71-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty