Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(8-Aminooctyl)-2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetamide 2,2,2-trifluoroacetate 98%

Supplier: AMBEED, INC
Warning

A724281-10MG A724281-1MG A724281-25MG A724281-5MG A724281-100MG A724281-50MG
76651-682EA 50 USD InStock
76651-682 76651-684 76651-686 76651-688 77622-586 77622-588
N-(8-Aminooctyl)-2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetamide 2,2,2-trifluoroacetate 98%
N-(8-Aminooctyl)-2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetamide 2,2,2-trifluoroacetate
Precaución: for Research Use Only


Formula: C₂₅H₃₁F₃N₄O₈
MW: 572.53 g/mol
MDL Number: MFCD31560485
CAS Number: 1950635-16-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty