Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(3-Bromo-4-methylphenyl)-4-(pyridin-4-ylmethyl)phthalazin-1-amine 98%

Supplier: AMBEED, INC
Warning

A725084-10MG A725084-1MG A725084-25MG A725084-50MG A725084-5MG
76651-802EA 25 USD InStock
76651-802 76651-804 76651-806 76651-808 76651-810
N-(3-Bromo-4-methylphenyl)-4-(pyridin-4-ylmethyl)phthalazin-1-amine 98%
N-(3-Bromo-4-methylphenyl)-4-(pyridin-4-ylmethyl)phthalazin-1-amine
Precaución: for Research Use Only


Formula: C₂₁H₁₇BrN₄
MW: 405.2905 g/mol
MDL Number: MFCD17014834
CAS Number: 300842-64-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty