Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N'-bis(2-Phenylphenyl)ethanediamide 98%

Supplier: AMBEED, INC
Danger

A725596-100G A725596-1G A725596-250MG A725596-25G A725596-5G
76651-858EA 531 USD InStock
76651-858 76651-860 76651-862 76651-864 76651-866
N,N'-bis(2-Phenylphenyl)ethanediamide 98%
N,N'-bis(2-Phenylphenyl)ethanediamide
Precaución: for Research Use Only


Formula: C₂₆H₂₀N₂O₂
MDL Number: MFCD31793560
CAS Number: 21022-17-3
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty