Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-(chloromethyl)bicyclo[1.1.1]pentane 98%

Supplier: AMBEED, INC
Danger

A728086-100MG A728086-250MG
76651-906EA 108 USD InStock
76651-906 76651-908
1-Bromo-3-(chloromethyl)bicyclo[1.1.1]pentane 98%
1-Bromo-3-(chloromethyl)bicyclo[1.1.1]pentane
Precaución: for Research Use Only


Formula: C₆H₈BrCl
MW: 195.48 g/mol
MDL Number: MFCD20624436
CAS Number: 137741-18-5
UN: 2920
ADR: 8,3 Ⅱ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty