Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-4-methoxy-L-homophenylalanine 95%

Supplier: AMBEED, INC
Warning

A729141-100MG A729141-50MG
76651-942EA 235 USD InStock
76651-942 76651-944
Fmoc-4-methoxy-L-homophenylalanine 95%
Fmoc-4-methoxy-L-homophenylalanine
Precaución: for Research Use Only


Formula: C₂₆H₂₅NO₅
MW: 431.4804 g/mol
MDL Number: MFCD01321419
CAS Number: 205182-52-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty