Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-(methoxymethyl)phenol 96%

Supplier: AMBEED, INC
Warning

A733149-100MG A733149-1G A733149-250MG
76652-016EA 19 USD InStock
76652-016 76652-018 76652-020
4-Fluoro-2-(methoxymethyl)phenol 96%
4-Fluoro-2-(methoxymethyl)phenol
Precaución: for Research Use Only


Formula: C₈H₉FO₂
MDL Number: MFCD31653804
CAS Number: 2222511-96-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty