Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Allyl-3-methylimidazolium tetrafluoroborate 99%

Supplier: AMBEED, INC
Danger

A736600-1G A736600-25G A736600-5G
76652-048EA 17 USD InStock
76652-048 76652-050 76652-052
1-Allyl-3-methylimidazolium tetrafluoroborate 99%
1-Allyl-3-methylimidazolium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₇H₁₁BF₄N₂
MDL Number: MFCD11109344
CAS Number: 851606-63-8
UN: 1760
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty