Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Carbamoyl-4-fluorobenzeneboronic acid pinacol ester 95%

Supplier: AMBEED, INC
Warning

A741619-100MG A741619-1G A741619-250MG A741619-5G
76652-118EA 39 USD InStock
76652-118 76652-120 76652-122 76652-124
3-Carbamoyl-4-fluorobenzeneboronic acid pinacol ester 95%
3-Carbamoyl-4-fluorobenzeneboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₇BFNO₃
MDL Number: MFCD18730402
CAS Number: 1374508-59-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty