Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boric acid, pinacol ester 98%

Supplier: AMBEED, INC
Warning

A742308-1G A742308-250MG A742308-25G A742308-5G
76652-144EA 83 USD InStock
76652-144 76652-146 76652-148 76652-150
Boric acid, pinacol ester 98%
Boric acid, pinacol ester
Precaución: for Research Use Only


Formula: C₆H₁₃BO₃
MDL Number: MFCD15143603
CAS Number: 25240-59-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty