Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 95%

Supplier: AMBEED, INC
Warning

A742360-100MG A742360-1G A742360-250MG
76652-152EA 39 USD InStock
76652-152 76652-154 76652-156
3-Amino-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 95%
3-Amino-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Precaución: for Research Use Only


Formula: C₁₃H₁₇BN₂O₂
MDL Number: MFCD22494384
CAS Number: 1036991-35-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty