Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Bromo-4-chlorophenyl)-2,2,2-trifluoroethanone 95%

Supplier: AMBEED, INC
Warning

A746690-1G A746690-250MG A746690-5G
76652-200EA 43 USD InStock
76652-200 76652-202 76652-204
1-(2-Bromo-4-chlorophenyl)-2,2,2-trifluoroethanone 95%
1-(2-Bromo-4-chlorophenyl)-2,2,2-trifluoroethanone
Precaución: for Research Use Only


Formula: C₈H₃BrClF₃O
MW: 287.4610 g/mol
MDL Number: MFCD27665766
CAS Number: 1033805-23-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty