Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris[5-(1,1-dimethylethyl)-2-(2-pyridinyl-κN)phenyl-κC]iridium 98%

Supplier: AMBEED, INC
Warning

A750571-100MG A750571-1G A750571-250MG
76652-332EA 55 USD InStock
76652-332 76652-334 76652-336
Tris[5-(1,1-dimethylethyl)-2-(2-pyridinyl-κN)phenyl-κC]iridium 98%
Tris[5-(1,1-dimethylethyl)-2-(2-pyridinyl-κN)phenyl-κC]iridium
Precaución: for Research Use Only


Formula: C₄₅H₄₈IrN₃
CAS Number: 359014-76-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty